Please login to your online account to display your discounted pricing

1-tert-Butyl 6-methyl 3-iodo-1H-indole-1,6-dicarboxylate, ≥97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-(tert-Butyl)-6-methyl-3-iodo-1H-indole-1, 6-dicarboxylate
CAS 850374-94-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C15H16INO4
Formula Weight 401.19
MDL Number MFCD06797499
Synonym 1-(tert-butyl) 6-methyl 3-iodo-1h-indole-1, 6-dicarboxylate (IUPAC)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO07419CB

  SDS
fisher scientific company
MO07419CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 401.19
Quantity 250mg
Assay 97%
CAS Min % 97.0
Molecular Formula C15H16INO4
CAS 850374-94-6
Synonym 1-(tert-butyl) 6-methyl 3-iodo-1h-indole-1, 6-dicarboxylate (IUPAC)
Packaging Amber Glass Bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 1-(tert-Butyl)-6-methyl-3-iodo-1H-indole-1, 6-dicarboxylate
MDL Number MFCD06797499
Assay Percent Range 97%