Please login to your online account to display your discounted pricing

1-tert-Butyl 6-methyl 3-iodo-1H-indole-1,6-dicarboxylate, ≥97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO07419CB

 View more versions of this product

Catalog No. MO07419CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 401.19
Quantity 250mg
Assay 97%
CAS Min % 97.0
Molecular Formula C15H16INO4
CAS 850374-94-6
Chemical Name or Material 1-(tert-Butyl)-6-methyl-3-iodo-1H-indole-1, 6-dicarboxylate
Assay Percent Range 97%
Packaging Amber Glass Bottle
MDL Number MFCD06797499
Synonym 1-(tert-butyl) 6-methyl 3-iodo-1h-indole-1, 6-dicarboxylate (IUPAC)
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-(tert-Butyl)-6-methyl-3-iodo-1H-indole-1, 6-dicarboxylate
CAS 850374-94-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C15H16INO4
Formula Weight 401.19
MDL Number MFCD06797499
Synonym 1-(tert-butyl) 6-methyl 3-iodo-1h-indole-1, 6-dicarboxylate (IUPAC)
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.