Please login to your online account to display your discounted pricing

1-(tert-butyl)-5-methyl-1H-pyrazole-3-carbohydrazide, Maybridge

Chemical Identifiers

Chemical Name or Material 1-(tert-Butyl)-5-methyl-1H-pyrazole-3-carbohydrazide
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H16N4O
Formula Weight 196.25
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB08431DA

 
fisher scientific company
SPB08431DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 196.25
Quantity 1g
Assay 97%
Assay Percent Range 97%
Chemical Name or Material 1-(tert-Butyl)-5-methyl-1H-pyrazole-3-carbohydrazide
Molecular Formula C9H16N4O
CAS 306937-23-5
Health Hazard 1 GHS Signal Word: Warning