Please login to your online account to display your discounted pricing

1-(tert-butyl)-5-methyl-1H-pyrazole-3-carbohydrazide, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  SPB08431DA

 View more versions of this product

Catalog No. SPB08431DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 196.25
Quantity 1g
Assay 97%
Assay Percent Range 97%
Chemical Name or Material 1-(tert-Butyl)-5-methyl-1H-pyrazole-3-carbohydrazide
CAS 306937-23-5
Molecular Formula C9H16N4O
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material 1-(tert-Butyl)-5-methyl-1H-pyrazole-3-carbohydrazide
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H16N4O
Formula Weight 196.25
Accessories

GHS Signal Word: Warning

SDS & Safety Documents