Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-tert-Butyl-4-nitrobenzene, 98%

$136.69 - $472.55

Chemical Identifiers

Chemical Name or Material 1-tert-Butyl-4-nitrobenzene
CAS 3282-56-2
Molecular Formula C10H13NO2
Formula Weight 179.22
MDL Number MFCD00041253
Beilstein 2047337
Synonym 4-Nitro-tert-butylbenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0606709

  SDS
alfa aesar™
L0606709
10g Each for $136.69

AAL0606718

  SDS
alfa aesar™
L0606718
50g Each for $472.55
Description & Specifications

Specifications

Formula Weight 179.22
Chemical Name or Material 1-tert-Butyl-4-nitrobenzene
Assay 98%
Molecular Formula C10H13NO2
MDL Number MFCD00041253
CAS 3282-56-2
Synonym 4-Nitro-tert-butylbenzene
Health Hazard 1 Danger
Beilstein 2047337
Health Hazard 2 GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 265° to 267°C
Quantity 10g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.