Please login to your online account to display your discounted pricing

1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carbonyl chloride, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carbonyl chloride
CAS 175277-10-8
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H13ClN2O
Formula Weight 200.67
MDL Number MFCD00084915
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB03129CB

  SDS
fisher scientific company
SPB03129CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 200.67
Quantity 250mg
Assay 95%
CAS Min % 95.0
Chemical Name or Material 1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carbonyl chloride
Molecular Formula C9H13ClN2O
MDL Number MFCD00084915
CAS 175277-10-8
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 95%
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.