Please login to your online account to display your discounted pricing

1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carbonyl chloride, 95%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  SPB03129CB

 View more versions of this product

Catalog No. SPB03129CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 200.67
Quantity 250mg
Assay 95%
CAS Min % 95.0
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carbonyl chloride
Molecular Formula C9H13ClN2O
CAS 175277-10-8
MDL Number MFCD00084915
Health Hazard 1 GHS Signal Word: Danger
Assay Percent Range 95%
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material 1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carbonyl chloride
CAS 175277-10-8
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H13ClN2O
Formula Weight 200.67
MDL Number MFCD00084915
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents