Please login to your online account to display your discounted pricing

1-(tert-Butyl)-3,5-dimethyl-1H-pyrazole-4-carbaldehyde, 97%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CD07394DA

 View more versions of this product

Catalog No. CD07394DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 180.25
Quantity 1g
Assay 97%
CAS Max % 100.0
Synonym 1-(tert-Butyl)-3, 5-dimethyl-1H-pyrazole-4-carbaldehyde (IUPAC)
Packaging Amber glass bottle
Molecular Formula C10H16N2O
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 96.5
CAS 647824-51-9
Chemical Name or Material 1-(Tert-butyl)-3, 5-dimethyl-1H-pyrazole-4-carbaldehyde
MDL Number MFCD00120293
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

Specifications

Chemical Name or Material 1-(Tert-butyl)-3, 5-dimethyl-1H-pyrazole-4-carbaldehyde
CAS 647824-51-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H16N2O
Formula Weight 180.25
MDL Number MFCD00120293
Synonym 1-(tert-Butyl)-3, 5-dimethyl-1H-pyrazole-4-carbaldehyde (IUPAC)
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents