Please login to your online account to display your discounted pricing

1-(tert-butyl) 2-methyl 1H-indole-1,2-dicarboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-(tert-Butyl) 2-methyl 1H-indole-1, 2-dicarboxylate
Assay 97%
Assay Percent Range 97%
Molecular Formula C15H17NO4
Formula Weight 275.3
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC27822CB

  SDS
fisher scientific company
CC27822CB
Amber glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC27822DE

  SDS
fisher scientific company
CC27822DE
Amber glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC27822DA

  SDS
fisher scientific company
CC27822DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 275.3
Quantity 250mg
Assay 97%
Chemical Name or Material 1-(tert-Butyl) 2-methyl 1H-indole-1, 2-dicarboxylate
Assay Percent Range 97%
Molecular Formula C15H17NO4
Packaging Amber glass bottle
CAS 163229-48-9