missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-tert-Butyl-2-imidazolidinone, 98+%

$265.24 - $265.24

Chemical Identifiers

CAS 92075-16-6
Molecular Formula C7H14N2O
Molecular Weight (g/mol) 142.202
MDL Number MFCD00832245
InChI Key VNXBETOLLOJKCA-UHFFFAOYSA-N
Synonym 1-tert-butyl-2-imidazolidinone, 1-tert-butyl imidazolidin-2-one, 1-tert-butyl-imidazolidin-2-one, 1-tert-butylimidazolidine-2-one, 1-tert-butyl-2-oxoimidazolidine, 1-tert-butyl tetrahydro-2h-imidazol-2-one
PubChem CID 2795231
IUPAC Name 1-tert-butylimidazolidin-2-one
SMILES CC(C)(C)N1CCNC1=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1762506 View Documents Alfa Aesar™
L1762506
5g Each for $265.24
 
Specifications

Chemical Identifiers

92075-16-6
142.202
VNXBETOLLOJKCA-UHFFFAOYSA-N
2795231
CC(C)(C)N1CCNC1=O
C7H14N2O
MFCD00832245
1-tert-butyl-2-imidazolidinone, 1-tert-butyl imidazolidin-2-one, 1-tert-butyl-imidazolidin-2-one, 1-tert-butylimidazolidine-2-one, 1-tert-butyl-2-oxoimidazolidine, 1-tert-butyl tetrahydro-2h-imidazol-2-one
1-tert-butylimidazolidin-2-one

Specifications

136°C to 138°C
92075-16-6
MFCD00832245
1-tert-butyl-2-imidazolidinone, 1-tert-butyl imidazolidin-2-one, 1-tert-butyl-imidazolidin-2-one, 1-tert-butylimidazolidine-2-one, 1-tert-butyl-2-oxoimidazolidine, 1-tert-butyl tetrahydro-2h-imidazol-2-one
CC(C)(C)N1CCNC1=O
142.202
142.2
1-tert-Butyl-2-imidazolidinone
5g
C7H14N2O
5499181
VNXBETOLLOJKCA-UHFFFAOYSA-N
1-tert-butylimidazolidin-2-one
2795231
≥98%