missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(tert-Butoxycarbonyl)-4-piperidone, TCI America™

$42.78 - $131.39

Chemical Identifiers

CAS 79099-07-3
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD00151800
InChI Key ROUYFJUVMYHXFJ-UHFFFAOYSA-N
Synonym 1-boc-4-piperidone, n-boc-4-piperidone, n-tert-butoxycarbonyl-4-piperidone, 1-boc-4-piperidinone, boc-4-piperidone, 1-tert-butoxycarbonyl-4-piperidone, n-tert-butoxycarbonyl-4-piperidone, n-boc-4-piperidinone, boc-piperidone, 1-t-boc-4-piperidone
PubChem CID 735900
IUPAC Name tert-butyl 4-oxopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(=O)CC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B23655G View Documents TCI America
B23655G
5g Each for $42.78
 
B236525G View Documents TCI America
B236525G
25g Each for $131.39
 
Specifications

Chemical Identifiers

79099-07-3
199.25
ROUYFJUVMYHXFJ-UHFFFAOYSA-N
735900
CC(C)(C)OC(=O)N1CCC(=O)CC1
C10H17NO3
MFCD00151800
1-boc-4-piperidone, n-boc-4-piperidone, n-tert-butoxycarbonyl-4-piperidone, 1-boc-4-piperidinone, boc-4-piperidone, 1-tert-butoxycarbonyl-4-piperidone, n-tert-butoxycarbonyl-4-piperidone, n-boc-4-piperidinone, boc-piperidone, 1-t-boc-4-piperidone
tert-butyl 4-oxopiperidine-1-carboxylate

Specifications

Undesignated
5g
C10H17NO3
1-boc-4-piperidone, n-boc-4-piperidone, n-tert-butoxycarbonyl-4-piperidone, 1-boc-4-piperidinone, boc-4-piperidone, 1-tert-butoxycarbonyl-4-piperidone, n-tert-butoxycarbonyl-4-piperidone, n-boc-4-piperidinone, boc-piperidone, 1-t-boc-4-piperidone
CC(C)(C)OC(=O)N1CCC(=O)CC1
199.25
199.25
≥98.0% (GC)
75°C
79099-07-3
MFCD00151800
ROUYFJUVMYHXFJ-UHFFFAOYSA-N
tert-butyl 4-oxopiperidine-1-carboxylate
735900
Crystal-Powder at 20°C
1-(tert-Butoxycarbonyl)-4-piperidone
Safety and Handling

Safety and Handling

TSCA : No