missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(tert-Butoxycarbonyl)-3-piperidinemethanol 95.0+%, TCI America™
Supplier: TCI America B378725G
Specifications
1-(tert-Butoxycarbonyl)-3-piperidinemethanol | |
78°C | |
C11H21NO3 | |
25 g | |
OJCLHERKFHHUTB-UHFFFAOYSA-N | |
tert-butyl 3-(hydroxymethyl)piperidine-1-carboxylate | |
2763851 | |
≥95.0% (GC) |
116574-71-1 | |
White-Yellow | |
MFCD03094733 | |
n-boc-piperidine-3-methanol, tert-butyl 3-hydroxymethyl piperidine-1-carboxylate, 1-boc-3-piperidinemethanol, n-boc-3-piperidinemethanol, 3-hydroxymethyl-1-n-boc-piperidine, n-boc-3-hydroxymethylpiperidine, 1-boc-3-hydroxymethylpiperidine, 1-boc-3-hydroxymethyl piperidine, tert-butyl 3-hydroxymethyl tetrahydro-1 2h-pyridinecarboxylate, +/--n-boc-3-piperidine methanol | |
CC(C)(C)OC(=O)N1CCCC(C1)CO | |
215.293 | |
215.29 | |
Crystalline Powder |
Chemical Identifiers
116574-71-1 | |
215.293 | |
OJCLHERKFHHUTB-UHFFFAOYSA-N | |
2763851 | |
CC(C)(C)OC(=O)N1CCCC(C1)CO |
C11H21NO3 | |
MFCD03094733 | |
n-boc-piperidine-3-methanol, tert-butyl 3-hydroxymethyl piperidine-1-carboxylate, 1-boc-3-piperidinemethanol, n-boc-3-piperidinemethanol, 3-hydroxymethyl-1-n-boc-piperidine, n-boc-3-hydroxymethylpiperidine, 1-boc-3-hydroxymethylpiperidine, 1-boc-3-hydroxymethyl piperidine, tert-butyl 3-hydroxymethyl tetrahydro-1 2h-pyridinecarboxylate, +/--n-boc-3-piperidine methanol | |
tert-butyl 3-(hydroxymethyl)piperidine-1-carboxylate |
Safety and Handling
TSCA : No