missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(tert-Butoxycarbonyl)-3-piperidinecarboxylic Acid, TCI America™

$65.70 - $195.56

Chemical Identifiers

CAS 84358-12-3
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD00673775
InChI Key NXILIHONWRXHFA-UHFFFAOYSA-N
Synonym 1-boc-piperidine-3-carboxylic acid, 1-tert-butoxycarbonyl piperidine-3-carboxylic acid, 1-boc-3-piperidinecarboxylic acid, n-boc-nipecotic acid, 1-tert-butoxycarbonyl-3-piperidinecarboxylic acid, boc-dl-nip-oh, n-boc-3-piperidinecarboxylic acid, n-boc-piperidine-3-carboxylic acid, 1-n-boc-piperidine-3-carboxylic acid, n-boc-dl-nipecotic acid
PubChem CID 2735647
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-3-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCC(C1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B33765G View Documents TCI America
B33765G
5g Each for $65.70
 
B337625G View Documents TCI America
B337625G
25g Each for $195.56
 
Specifications

Chemical Identifiers

84358-12-3
229.276
NXILIHONWRXHFA-UHFFFAOYSA-N
2735647
CC(C)(C)OC(=O)N1CCCC(C1)C(=O)O
C11H19NO4
MFCD00673775
1-boc-piperidine-3-carboxylic acid, 1-tert-butoxycarbonyl piperidine-3-carboxylic acid, 1-boc-3-piperidinecarboxylic acid, n-boc-nipecotic acid, 1-tert-butoxycarbonyl-3-piperidinecarboxylic acid, boc-dl-nip-oh, n-boc-3-piperidinecarboxylic acid, n-boc-piperidine-3-carboxylic acid, 1-n-boc-piperidine-3-carboxylic acid, n-boc-dl-nipecotic acid
1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-3-carboxylic acid

Specifications

White-Yellow
5g
C11H19NO4
1-boc-piperidine-3-carboxylic acid, 1-tert-butoxycarbonyl piperidine-3-carboxylic acid, 1-boc-3-piperidinecarboxylic acid, n-boc-nipecotic acid, 1-tert-butoxycarbonyl-3-piperidinecarboxylic acid, boc-dl-nip-oh, n-boc-3-piperidinecarboxylic acid, n-boc-piperidine-3-carboxylic acid, 1-n-boc-piperidine-3-carboxylic acid, n-boc-dl-nipecotic acid
CC(C)(C)OC(=O)N1CCCC(C1)C(=O)O
229.276
229.28
≥97.0% (GC,T)
160°C
84358-12-3
MFCD00673775
NXILIHONWRXHFA-UHFFFAOYSA-N
1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-3-carboxylic acid
2735647
Crystal-Powder at 20°C
1-(tert-Butoxycarbonyl)-3-piperidinecarboxylic Acid
Safety and Handling

Safety and Handling

TSCA : No