missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(tert-Butoxycarbonyl)-3-fluoro-4-piperidone, TCI America™

$134.45 - $401.81

Chemical Identifiers

CAS 211108-50-8
Molecular Formula C10H16FNO3
Molecular Weight (g/mol) 217.24
MDL Number MFCD10566554
InChI Key JZNWQLLPLOQGOI-UHFFFAOYSA-N
Synonym 1-boc-3-fluoro-4-oxopiperidine, 3-fluoro-4-oxopiperidine-1-carboxylic acid tert-butyl ester, 1-boc-3-fluoro-4-piperidone, 1-tert-butoxycarbonyl-3-fluoro-4-piperidone, 1-piperidinecarboxylic acid, 3-fluoro-4-oxo-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-fluoro-4-oxopiperidine, 3-fluoro-4-oxo-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl3-fluoro-4-oxopiperidine-1-carboxylate, tert-butyl 3-fluoro-4-oxo-piperidine-1-carboxylate, n-boc-3-fluoro-4-piperidinone
PubChem CID 10560711
IUPAC Name tert-butyl 3-fluoro-4-oxopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(=O)C(C1)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B4858200MG View Documents TCI America
B4858200MG
200mg Each for $134.45
 
B48581G View Documents TCI America
B48581G
1g Each for $401.81
 
Specifications

Chemical Identifiers

211108-50-8
217.24
JZNWQLLPLOQGOI-UHFFFAOYSA-N
10560711
CC(C)(C)OC(=O)N1CCC(=O)C(C1)F
C10H16FNO3
MFCD10566554
1-boc-3-fluoro-4-oxopiperidine, 3-fluoro-4-oxopiperidine-1-carboxylic acid tert-butyl ester, 1-boc-3-fluoro-4-piperidone, 1-tert-butoxycarbonyl-3-fluoro-4-piperidone, 1-piperidinecarboxylic acid, 3-fluoro-4-oxo-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-fluoro-4-oxopiperidine, 3-fluoro-4-oxo-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl3-fluoro-4-oxopiperidine-1-carboxylate, tert-butyl 3-fluoro-4-oxo-piperidine-1-carboxylate, n-boc-3-fluoro-4-piperidinone
tert-butyl 3-fluoro-4-oxopiperidine-1-carboxylate

Specifications

White-Yellow
200mg
C10H16FNO3
1-boc-3-fluoro-4-oxopiperidine, 3-fluoro-4-oxopiperidine-1-carboxylic acid tert-butyl ester, 1-boc-3-fluoro-4-piperidone, 1-tert-butoxycarbonyl-3-fluoro-4-piperidone, 1-piperidinecarboxylic acid, 3-fluoro-4-oxo-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-fluoro-4-oxopiperidine, 3-fluoro-4-oxo-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl3-fluoro-4-oxopiperidine-1-carboxylate, tert-butyl 3-fluoro-4-oxo-piperidine-1-carboxylate, n-boc-3-fluoro-4-piperidinone
CC(C)(C)OC(=O)N1CCC(=O)C(C1)F
217.24
217.24
≥97.0% (GC)
77°C
211108-50-8
MFCD10566554
JZNWQLLPLOQGOI-UHFFFAOYSA-N
tert-butyl 3-fluoro-4-oxopiperidine-1-carboxylate
10560711
Crystal-Powder at 20°C
1-(tert-Butoxycarbonyl)-3-fluoro-4-piperidone
Safety and Handling

Safety and Handling

TSCA : No