missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-tert-Butoxycarbonyl-2-pyrrolylboronic acid, 98%, ACROS Organics™
Chemical Identifiers
CAS | 135884-31-0 |
---|---|
Molecular Formula | C9H14BNO4 |
Molecular Weight (g/mol) | 211.02 |
MDL Number | MFCD01318939 |
InChI Key | ZWGMJLNXIVRFRJ-UHFFFAOYSA-N |
Synonym | 1-boc-2-pyrrolylboronic acid, 1-boc-pyrrole-2-boronic acid, 1-t-butoxycarbonyl pyrrole-2-boronic acid, 1-tert-butoxy carbonyl-1h-pyrrol-2-yl boronic acid, 1-tert-butoxycarbonyl pyrrole-2-boronic acid, 1-tert-butoxycarbonyl-1h-pyrrol-2-yl boronic acid, 1-tert-butoxycarbonyl-1h-pyrrol-2-ylboronic acid, boc-pyrrole-2-boronic acid, n-boc-2-pyrroleboronic acid, n-boc-pyrrole-2-boronic acid |
PubChem CID | 2734321 |
IUPAC Name | [1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrol-2-yl]boronic acid |
SMILES | B(C1=CC=CN1C(=O)OC(C)(C)C)(O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC358982500
|
Thermo Fisher Scientific
358982500 |
250mg | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
135884-31-0 | |
211.02 | |
ZWGMJLNXIVRFRJ-UHFFFAOYSA-N | |
2734321 | |
B(C1=CC=CN1C(=O)OC(C)(C)C)(O)O |
C9H14BNO4 | |
MFCD01318939 | |
1-boc-2-pyrrolylboronic acid, 1-boc-pyrrole-2-boronic acid, 1-t-butoxycarbonyl pyrrole-2-boronic acid, 1-tert-butoxy carbonyl-1h-pyrrol-2-yl boronic acid, 1-tert-butoxycarbonyl pyrrole-2-boronic acid, 1-tert-butoxycarbonyl-1h-pyrrol-2-yl boronic acid, 1-tert-butoxycarbonyl-1h-pyrrol-2-ylboronic acid, boc-pyrrole-2-boronic acid, n-boc-2-pyrroleboronic acid, n-boc-pyrrole-2-boronic acid | |
[1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrol-2-yl]boronic acid |
Specifications
Authentic | |
Beige to Brown | |
135884-31-0 | |
100.0 | |
C9H14BNO4 | |
1-boc-2-pyrrolylboronic acid, 1-boc-pyrrole-2-boronic acid, 1-t-butoxycarbonyl pyrrole-2-boronic acid, 1-tert-butoxy carbonyl-1h-pyrrol-2-yl boronic acid, 1-tert-butoxycarbonyl pyrrole-2-boronic acid, 1-tert-butoxycarbonyl-1h-pyrrol-2-yl boronic acid, 1-tert-butoxycarbonyl-1h-pyrrol-2-ylboronic acid, boc-pyrrole-2-boronic acid, n-boc-2-pyrroleboronic acid, n-boc-pyrrole-2-boronic acid | |
B(C1=CC=CN1C(=O)OC(C)(C)C)(O)O | |
211.02 | |
211.02 | |
98% |
Glass bottle | |
250mg | |
97.5 | |
97.5% min. (HPLC) | |
MFCD01318939 | |
ZWGMJLNXIVRFRJ-UHFFFAOYSA-N | |
[1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrol-2-yl]boronic acid | |
2734321 | |
Crystalline Powder | |
1-tert-Butoxycarbonyl-2-pyrrolylboronic acid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
TSCA : TSCA