missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-tert-Butoxycarbonyl-2-pyrrolylboronic acid, 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 35898-2500

 View more versions of this product

Catalog No. AC358982500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

1-tert-Butoxycarbonyl-2-pyrrolylboronic acid
97.5
Authentic
≥97.5%
Glass bottle
211.02
Beige to light brown
135884-31-0
100.0
Crystalline Powder
98%
C9H14BNO4
MFCD01318939
250mg

Chemical Identifiers

135884-31-0
100.0
C9H14BNO4
MFCD01318939
≥97.5%
97.5
98%
211.02
1-tert-Butoxycarbonyl-2-pyrrolylboronic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA