missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-S-Octyl-β-D-thioglucopyranoside, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC315000100

 View more versions of this product

Catalog No. AC315000100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Powder
≥97.5% (HPLC)
White
100.0
97.5
Glass bottle
98%
1-S-Octyl-β-D-thioglucopyranoside
MFCD00012189
308.44
10g
− 46.00 (20.00°C c=1,CH3OH)
Authentic
−52° to −54° (20°C, 589nm) (c=1, CH3OH)
98%
C14H28O5S
85618-21-9

Chemical Identifiers

85618-21-9
100.0
C14H28O5S
MFCD00012189
≥97.5% (HPLC)
97.5
98%
308.44
1-S-Octyl-β-D-thioglucopyranoside, 98%

Safety and Handling

SDS & Safety Documents

TSCA : TSCA