Learn More
1-Pyrrolidino-1-cyclohexene, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 334831000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-Pyrrolidino-1-cyclohexene | |
1125-99-1 | |
100.0 | |
114°C to 115°C (15.0mmHg) | |
Authentic | |
Glass bottle | |
1.5210 to 1.523 | |
100g | |
1-pyrrolidino-1-cyclohexene, 1-pyrrolidinocyclohexene, 1-cyclohex-1-en-1-yl pyrrolidine, pyrrolidine, 1-1-cyclohexen-1-yl, 1-1-cyclohexen-1-yl pyrrolidine, 1-1-pyrrolidinyl cyclohexene, cyclohexanone pyrrolidine enamine, 1-pyrrolidinyl-1-cyclohexene, n-1-cyclohexenyl pyrrolidine, 1-1-cyclohexenyl pyrrolidine | |
C1CCC(=CC1)N2CCCC2 | |
151.25 | |
151.25 |
95% | |
94.0 | |
0.9400g/mL | |
66°C | |
94% min. (GC) | |
C10H17N | |
MFCD00003163 | |
0.94 | |
KTZNVZJECQAMBV-UHFFFAOYSA-N | |
1-(cyclohexen-1-yl)pyrrolidine | |
70768 | |
95% |
Chemical Identifiers
1125-99-1 | |
151.25 | |
KTZNVZJECQAMBV-UHFFFAOYSA-N | |
70768 | |
C1CCC(=CC1)N2CCCC2 |
C10H17N | |
MFCD00003163 | |
1-pyrrolidino-1-cyclohexene, 1-pyrrolidinocyclohexene, 1-cyclohex-1-en-1-yl pyrrolidine, pyrrolidine, 1-1-cyclohexen-1-yl, 1-1-cyclohexen-1-yl pyrrolidine, 1-1-pyrrolidinyl cyclohexene, cyclohexanone pyrrolidine enamine, 1-pyrrolidinyl-1-cyclohexene, n-1-cyclohexenyl pyrrolidine, 1-1-cyclohexenyl pyrrolidine | |
1-(cyclohexen-1-yl)pyrrolidine |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and
GHS Signal Word: Warning
EINECSNumber : 214-414-6
TSCA : TSCA
RUO â Research Use Only