missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Pyrrolidino-1-cyclohexene, 95%, Thermo Scientific™
Chemical Identifiers
CAS | 1125-99-1 |
---|---|
Molecular Formula | C10H17N |
Molecular Weight (g/mol) | 151.25 |
MDL Number | MFCD00003163 |
InChI Key | KTZNVZJECQAMBV-UHFFFAOYSA-N |
Synonym | 1-pyrrolidino-1-cyclohexene, 1-pyrrolidinocyclohexene, 1-cyclohex-1-en-1-yl pyrrolidine, pyrrolidine, 1-1-cyclohexen-1-yl, 1-1-cyclohexen-1-yl pyrrolidine, 1-1-pyrrolidinyl cyclohexene, cyclohexanone pyrrolidine enamine, 1-pyrrolidinyl-1-cyclohexene, n-1-cyclohexenyl pyrrolidine, 1-1-cyclohexenyl pyrrolidine |
PubChem CID | 70768 |
IUPAC Name | 1-(cyclohexen-1-yl)pyrrolidine |
SMILES | C1CCC(=CC1)N2CCCC2 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC334830250
|
Thermo Scientific Chemicals
334830250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC334831000
|
Thermo Scientific Chemicals
334831000 |
100g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1125-99-1 | |
151.25 | |
KTZNVZJECQAMBV-UHFFFAOYSA-N | |
70768 | |
C1CCC(=CC1)N2CCCC2 |
C10H17N | |
MFCD00003163 | |
1-pyrrolidino-1-cyclohexene, 1-pyrrolidinocyclohexene, 1-cyclohex-1-en-1-yl pyrrolidine, pyrrolidine, 1-1-cyclohexen-1-yl, 1-1-cyclohexen-1-yl pyrrolidine, 1-1-pyrrolidinyl cyclohexene, cyclohexanone pyrrolidine enamine, 1-pyrrolidinyl-1-cyclohexene, n-1-cyclohexenyl pyrrolidine, 1-1-cyclohexenyl pyrrolidine | |
1-(cyclohexen-1-yl)pyrrolidine |
Specifications
1125-99-1 | |
100.0 | |
114°C to 115°C (15.0mmHg) | |
Authentic | |
Glass bottle | |
1.5210 to 1.523 | |
25g | |
1-pyrrolidino-1-cyclohexene, 1-pyrrolidinocyclohexene, 1-cyclohex-1-en-1-yl pyrrolidine, pyrrolidine, 1-1-cyclohexen-1-yl, 1-1-cyclohexen-1-yl pyrrolidine, 1-1-pyrrolidinyl cyclohexene, cyclohexanone pyrrolidine enamine, 1-pyrrolidinyl-1-cyclohexene, n-1-cyclohexenyl pyrrolidine, 1-1-cyclohexenyl pyrrolidine | |
C1CCC(=CC1)N2CCCC2 | |
151.25 | |
151.25 | |
1-Pyrrolidino-1-cyclohexene, 95% |
94.0 | |
0.9400g/mL | |
66°C | |
94% min. (GC) | |
C10H17N | |
MFCD00003163 | |
0.94 | |
KTZNVZJECQAMBV-UHFFFAOYSA-N | |
1-(cyclohexen-1-yl)pyrrolidine | |
70768 | |
95% |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and
GHS Signal Word: Warning
EINECSNumber : 214-414-6
TSCA : TSCA
RUO â Research Use Only