missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Pyrrolidinepropionitrile, 97%

$55.51 - $178.89

Chemical Identifiers

CAS 26165-45-7
Molecular Formula C7H12N2
Molecular Weight (g/mol) 124.187
MDL Number MFCD00014099
InChI Key PDVYZQGNSCSKPG-UHFFFAOYSA-N
Synonym 1-pyrrolidinepropionitrile, 3-pyrrolidin-1-yl propanenitrile, 3-1-pyrrolidino propionitrile, 1-pyrrolidinepropanenitrile, beta-pyrrolidinopropionitrile, n-2-cyanoethyl pyrrolidine, pyrrolidine-1-propiononitrile, n-beta-cyanoethyl pyrrolidine, 3-1-pyrrolidine propionitrile, 3-1-pyrrolidinyl propionitrile
PubChem CID 117743
IUPAC Name 3-pyrrolidin-1-ylpropanenitrile
SMILES C1CCN(C1)CCC#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0181806 View Documents Alfa Aesar™
L0181806
5g Each for $55.51
 
AAL0181814 View Documents Alfa Aesar™
L0181814
25g Each for $178.89
 
Specifications

Chemical Identifiers

26165-45-7
124.187
PDVYZQGNSCSKPG-UHFFFAOYSA-N
117743
C1CCN(C1)CCC#N
C7H12N2
MFCD00014099
1-pyrrolidinepropionitrile, 3-pyrrolidin-1-yl propanenitrile, 3-1-pyrrolidino propionitrile, 1-pyrrolidinepropanenitrile, beta-pyrrolidinopropionitrile, n-2-cyanoethyl pyrrolidine, pyrrolidine-1-propiononitrile, n-beta-cyanoethyl pyrrolidine, 3-1-pyrrolidine propionitrile, 3-1-pyrrolidinyl propionitrile
3-pyrrolidin-1-ylpropanenitrile

Specifications

104°C to 105°C (15mmHg)
26165-45-7
MFCD00014099
107542
PDVYZQGNSCSKPG-UHFFFAOYSA-N
3-pyrrolidin-1-ylpropanenitrile
117743
97%
5g
C7H12N2
UN3276
1-pyrrolidinepropionitrile, 3-pyrrolidin-1-yl propanenitrile, 3-1-pyrrolidino propionitrile, 1-pyrrolidinepropanenitrile, beta-pyrrolidinopropionitrile, n-2-cyanoethyl pyrrolidine, pyrrolidine-1-propiononitrile, n-beta-cyanoethyl pyrrolidine, 3-1-pyrrolidine propionitrile, 3-1-pyrrolidinyl propionitrile
C1CCN(C1)CCC#N
124.187
124.19
1-Pyrrolidinepropionitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

RTECSNumber : UY2273000