Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Pyrrolidinecarbonyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 1-Pyrrolidinecarbonyl chloride
CAS 1192-63-8
Molecular Formula C5H8ClNO
Formula Weight 133.58
MDL Number MFCD00051321
UN Number UN3265
Beilstein 110281
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1339603

 
alfa aesar™
L1339603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1339606

 
alfa aesar™
L1339606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 133.58
Chemical Name or Material 1-Pyrrolidinecarbonyl chloride
Assay 97%
Molecular Formula C5H8ClNO
MDL Number MFCD00051321
CAS 1192-63-8
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
TSCA TSCA
UN Number UN3265
Beilstein 110281
Boiling Point 75°C (1mmHg)
Flash Point 97°C (206°F)
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Applications
1-Pyrrolidinecarbonyl chloride, is used in the synthesis of (S)-4-(2-(((benzyloxy)carbonyl)amino)-3-methoxy-3-oxopropyl)phenyl pyrrolidine-1-carboxylate

Solubility
It reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, bases. Stable under recommended storage conditions.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C