Learn More
1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%, Thermo Scientific™
1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%, C5H12N2S2, CAS Number-5108-96-3
Supplier: Thermo Scientific Chemicals 162055000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-Pyrrolidinecarbodithioic acid, ammonium salt | |
97.5 | |
Authentic | |
Glass bottle | |
MFCD00012720 | |
ammonium pyrrolidyldithiocarbamate, apdc, ammonium 1-pyrrolidinecarbodithioate, ammonium tetramethylenedithiocarbamate, ammonium pyrrolidine-1-carbodithioate, ammonium 1-pyrrolidinyldithiocarbamate, ammonium 1-pyrrolidinedithiocarboxylate, 1-pyrrolidinecarbodithioic acid, ammonium salt, 1-pyrrolidinecarbodithioic acid ammonium salt, pyrrolidinedithiocarbamate ammonium | |
VSWDORGPIHIGNW-UHFFFAOYSA-M | |
pyrrolidine-1-carbodithioate | |
4311638 | |
98% |
5108-96-3 | |
100.0 | |
98% | |
C5H12N2S2 | |
500g | |
Solubility in water: 50g/L (20°C). Other solubilities: soluble in ethanol and chloroform | |
C1CCN(C1)C(=S)[S-] | |
164.28 | |
164.28 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap a
GHS Signal Word: Warning
EINECSNumber : 225-834-4
RUO â Research Use Only