missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%, ACROS Organics™

1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%, C5H12N2S2, CAS Number-5108-96-3

Manufacturer:  Thermo Fisher Scientific 162051000

 View more versions of this product

Catalog No. AC162051000



Description

Description

1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%, C5H12N2S2, CAS Number-5108-96-3

Specifications

Specifications

1-Pyrrolidinecarbodithioic acid, ammonium salt
Glass bottle
150.0°C to 155.0°C
5108-96-3
100.0
C5H12N2S2
1-pyrrolidinecarbodithioic acid ammonium salt, 1-pyrrolidinecarbodithioic acid, ammonium salt, ammonium 1-pyrrolidinecarbodithioate, ammonium 1-pyrrolidinedithiocarboxylate, ammonium 1-pyrrolidinyldithiocarbamate, ammonium pyrrolidine-1-carbodithioate, ammonium pyrrolidyldithiocarbamate, ammonium tetramethylenedithiocarbamate, apdc, pyrrolidinedithiocarbamate ammonium
VSWDORGPIHIGNW-UHFFFAOYSA-M
pyrrolidine-1-carbodithioate
4311638
Powder
Authentic
White
100g
97.5
98%
MFCD00012720
Solubility in water: 50g/L (20°C). Other solubilities: soluble in ethanol and chloroform
C1CCN(C1)C(=S)[S-]
164.28
164.28
98%

Chemical Identifiers

5108-96-3
164.28
VSWDORGPIHIGNW-UHFFFAOYSA-M
4311638
C1CCN(C1)C(=S)[S-]
C5H12N2S2
MFCD00012720
1-pyrrolidinecarbodithioic acid ammonium salt, 1-pyrrolidinecarbodithioic acid, ammonium salt, ammonium 1-pyrrolidinecarbodithioate, ammonium 1-pyrrolidinedithiocarboxylate, ammonium 1-pyrrolidinyldithiocarbamate, ammonium pyrrolidine-1-carbodithioate, ammonium pyrrolidyldithiocarbamate, ammonium tetramethylenedithiocarbamate, apdc, pyrrolidinedithiocarbamate ammonium
pyrrolidine-1-carbodithioate
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap a

GHS Signal Word: Warning

EINECSNumber : 225-834-4

SDS Documents