missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Pyrrolidinebutanenitrile, 97%, ACROS Organics™

Chemical Identifiers

CAS 35543-25-0
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H14N2
Formula Weight 138.21
MDL Number MFCD00003183
Synonym 1-Pyrrolidinobutyronitrile
Chemical Name or Material 1-Pyrrolidinebutanenitrile, 97%
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC123330250 SDS Thermo Fisher Scientific
123330250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

35543-25-0
100.0
C8H14N2
MFCD00003183
1-Pyrrolidinebutanenitrile, 97%
96.0
97%
138.21
1-Pyrrolidinobutyronitrile
≥96.0%

Specifications

25g
96.0
Authentic
1.4600 to 1.4620 (20°C, 589nm)
97%
138.21
0.9200g/mL
Clear colorless to brown
≥96.0%
35543-25-0
100.0
Glass bottle
Liquid
C8H14N2
MFCD00003183
1-Pyrrolidinobutyronitrile
1-Pyrrolidinebutanenitrile, 97%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 252-612-4

RTECSNumber : UY0270000

TSCA : TSCA