missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Pyrimidin-2-yl-piperidine-4-carboxylic acid, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC20701DA
Specifications
1-Pyrimidin-2-yl-piperidine-4-carboxylic acid | |
Amber glass bottle | |
MFCD00664474 | |
1-pyrimidin-2-yl piperidine-4-carboxylic acid, 1-pyrimidin-2-yl-piperidine-4-carboxylic acid, 1-2-pyrimidinyl piperidine-4-carboxylic acid, 1-2-pyrimidinyl-4-piperidinecarboxylic acid, 1-2-pyrimidyl piperidine-4-carboxylic acid, 4-piperidinecarboxylic acid, 1-2-pyrimidinyl, acmc-1agno, 4-piperidinecarboxylicacid, 1-2-pyrimidinyl | |
[O-]C(=O)C1CCN(CC1)C1=NC=CC=N1 | |
206.23 | |
207.23 |
303144-44-7 | |
C10H12N3O2 | |
1g | |
DYVXURZASBPYFR-UHFFFAOYSA-M | |
1-pyrimidin-2-ylpiperidine-4-carboxylic acid | |
2767267 | |
97% |
Chemical Identifiers
303144-44-7 | |
206.23 | |
DYVXURZASBPYFR-UHFFFAOYSA-M | |
2767267 | |
[O-]C(=O)C1CCN(CC1)C1=NC=CC=N1 |
C10H12N3O2 | |
MFCD00664474 | |
1-pyrimidin-2-yl piperidine-4-carboxylic acid, 1-pyrimidin-2-yl-piperidine-4-carboxylic acid, 1-2-pyrimidinyl piperidine-4-carboxylic acid, 1-2-pyrimidinyl-4-piperidinecarboxylic acid, 1-2-pyrimidyl piperidine-4-carboxylic acid, 4-piperidinecarboxylic acid, 1-2-pyrimidinyl, acmc-1agno, 4-piperidinecarboxylicacid, 1-2-pyrimidinyl | |
1-pyrimidin-2-ylpiperidine-4-carboxylic acid |