missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Pyrenecarboxaldehyde, 99%

Manufacturer:  Alfa Aesar™ L1083514

 View more versions of this product

Catalog No. AAL1083514



Description

Description

Applications
1-Pyrenecarboxaldehyde is a important intermediate used in agrochemical, pharmaceutical and dyestuff field. It is used as a highly polarity-sensitive fluorophore for SNP typing.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
Specifications

Specifications

1-Pyrenecarboxaldehyde
25g
C17H10O
1874889
RCYFOPUXRMOLQM-UHFFFAOYSA-N
pyrene-1-carbaldehyde
232848
99%
125°C to 128°C
3029-19-4
MFCD00004139
1-pyrenecarboxaldehyde, 1-formylpyrene, 1-pyrenealdehyde, pyrenecarboxaldehyde, 3-formylpyrene, 3-pyrenealdehyde, 3-pyrenylaldehyde, pyrene-1-aldehyde, 3-pyrenecarboxaldehyde, 1-pyrenecarbaldehyde
C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)C=O
230.266
230.27

Chemical Identifiers

3029-19-4
230.266
RCYFOPUXRMOLQM-UHFFFAOYSA-N
232848
C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)C=O
C17H10O
MFCD00004139
1-pyrenecarboxaldehyde, 1-formylpyrene, 1-pyrenealdehyde, pyrenecarboxaldehyde, 3-formylpyrene, 3-pyrenealdehyde, 3-pyrenylaldehyde, pyrene-1-aldehyde, 3-pyrenecarboxaldehyde, 1-pyrenecarbaldehyde
pyrene-1-carbaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UR2450200

Documents