Please login to your online account to display your discounted pricing

1-Propynylmagnesium bromide, 0.5M solution in THF, AcroSeal™, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass bottle

Manufacturer: thermo fisher scientific  AC426078000

 View more versions of this product

Catalog No. AC426078000

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material 1-Propynylmagnesium bromide
Quantity 800mL
CAS 16466-97-0
CAS 109-99-9
Packaging AcroSeal™ Glass bottle
Molecular Formula C3H3BrMg
Formula Weight 143.26
MDL Number MFCD00216588

Specifications

Chemical Name or Material 1-Propynylmagnesium bromide
CAS 16466-97-0
CAS 109-99-9
Molecular Formula C3H3BrMg
Formula Weight 143.26
MDL Number MFCD00216588