Learn More
1-Propylphosphonic acid cyclic anhydride, 50+% w/w soln. in dichloromethane, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6358330
Description
1-Propylphosphonic acid cyclic anhydride is used as a reagent for amide/peptide bond formation. It is utilized for condensation reactions and alcohol oxidations. It is also used to promote the synthesis of hydroxamic acids from carboxylic acids. Further, it is employed as an efficient promoter for the Lossen rearrangement to synthesis urea and carbamate derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Propylphosphonic acid cyclic anhydride is used as a reagent for amide/peptide bond formation. It is utilized for condensation reactions and alcohol oxidations. It is also used to promote the synthesis of hydroxamic acids from carboxylic acids. Further, it is employed as an efficient promoter for the Lossen rearrangement to synthesis urea and carbamate derivatives.
Solubility
Miscible with dioxane, terahydrofuran, dimethyl formamide, polar and aprotic organic solvents.
Notes
Moisture sensitive. Incompatible withstrong oxidizing agents.
Specifications
1-Propylphosphonic acid cyclic anhydride | |
C9H21O6P3 | |
250 g | |
5079255 | |
Miscible with dioxane,terahydrofuran,dimethyl formamide,polar and aprotic organic solvents. | |
CCCP1(=O)OP(=O)(OP(=O)(O1)CCC)CCC | |
318.182 | |
318.19 |
68957-94-8 | |
MFCD00006583 | |
UN2922 | |
2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphorinane-2,4,6-trioxide, t3p, 2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphinane 2,4,6-trioxide, unii-i6egd8839n, unii-i6egd8839n, 1,3,5,2,4,6-trioxatriphosphorinane, 2,4,6-tripropyl-, 2,4,6-trioxide, 2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphorinane 2,4,6-trioxide, 1-propanephosphonic anhydride in thf, 2,4,6-tri-n-propyl-2,4,6-trioxo-1,3,5,2,4,6-trioxatriphosphorinane, 1-propanephosphonic anhydride in toluene, 1-propanephosphonic anhydride in toluene | |
PAQZWJGSJMLPMG-UHFFFAOYSA-N | |
2,4,6-tripropyl-1,3,5,2$l^{5},4$l^{5},6$l^{5}-trioxatriphosphinane 2,4,6-trioxide | |
111923 | |
≥50% w/w soln. in dichloromethane |
Chemical Identifiers
68957-94-8 | |
318.182 | |
PAQZWJGSJMLPMG-UHFFFAOYSA-N | |
111923 | |
CCCP1(=O)OP(=O)(OP(=O)(O1)CCC)CCC |
C9H21O6P3 | |
MFCD00006583 | |
2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphorinane-2,4,6-trioxide, t3p, 2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphinane 2,4,6-trioxide, unii-i6egd8839n, unii-i6egd8839n, 1,3,5,2,4,6-trioxatriphosphorinane, 2,4,6-tripropyl-, 2,4,6-trioxide, 2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphorinane 2,4,6-trioxide, 1-propanephosphonic anhydride in thf, 2,4,6-tri-n-propyl-2,4,6-trioxo-1,3,5,2,4,6-trioxatriphosphorinane, 1-propanephosphonic anhydride in toluene, 1-propanephosphonic anhydride in toluene | |
2,4,6-tripropyl-1,3,5,2$l^{5},4$l^{5},6$l^{5}-trioxatriphosphinane 2,4,6-trioxide |
Safety and Handling
GHS H Statement
H314-H351
Causes severe skin burns and eye damage.
Suspected of causing cancer.
P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335-H336-H350
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 422-210-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only