missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Propylphosphonic acid cyclic anhydride, 50+% w/w soln. in acetonitrile

Manufacturer:  Alfa Aesar™ H6696730

 View more versions of this product

Catalog No. AAH6696730



Description

Description

Applications
1-Propylphosphonic acid cyclic anhydride is used as a reagent for amide/peptide bond formation. It is utilized for condensation reactions and alcohol oxidations. It is also used to promote the synthesis of hydroxamic acids from carboxylic acids. Further, it is employed as an efficient promoter for the Lossen rearrangement to synthesis urea and carbamate derivatives.

Solubility
Miscible with dioxane, terahydrofuran, dimethyl formamide, polar and aprotic organic solvents.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
Specifications

Specifications

250g
C9H21O6P3
UN2924
1,3,5,2,4,6-trioxatriphosphorinane, 2,4,6-tripropyl-, 2,4,6-trioxide, 1-propanephosphonic acid cyclic anhydride, 1-propanephosphonic anhydride in thf, 1-propanephosphonic anhydride in toluene, 2,4,6-tri-n-propyl-2,4,6-trioxo-1,3,5,2,4,6-trioxatriphosphorinane, 2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphinane 2,4,6-trioxide, 2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphorinane 2,4,6-trioxide, 2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphorinane-2,4,6-trioxide, propane phosphonic acid anhydride, unii-i6egd8839n
Miscible with dioxane,terahydrofuran,dimethyl formamide,polar and aprotic organic solvents.
CCCP1(=O)OP(=O)(OP(=O)(O1)CCC)CCC
318.182
318.19
68957-94-8
MFCD00006583
5079255
≥50% w/w soln. in acetonitrile
PAQZWJGSJMLPMG-UHFFFAOYSA-N
2,4,6-tripropyl-1,3,5,2$l^{5},4$l^{5},6$l^{5}-trioxatriphosphinane 2,4,6-trioxide
111923

Chemical Identifiers

68957-94-8
318.182
PAQZWJGSJMLPMG-UHFFFAOYSA-N
111923
CCCP1(=O)OP(=O)(OP(=O)(O1)CCC)CCC
C9H21O6P3
MFCD00006583
1,3,5,2,4,6-trioxatriphosphorinane, 2,4,6-tripropyl-, 2,4,6-trioxide, 1-propanephosphonic acid cyclic anhydride, 1-propanephosphonic anhydride in thf, 1-propanephosphonic anhydride in toluene, 2,4,6-tri-n-propyl-2,4,6-trioxo-1,3,5,2,4,6-trioxatriphosphorinane, 2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphinane 2,4,6-trioxide, 2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphorinane 2,4,6-trioxide, 2,4,6-tripropyl-1,3,5,2,4,6-trioxatriphosphorinane-2,4,6-trioxide, propane phosphonic acid anhydride, unii-i6egd8839n
2,4,6-tripropyl-1,3,5,2$l^{5},4$l^{5},6$l^{5}-trioxatriphosphinane 2,4,6-trioxide
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H312
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful in contact with skin.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302+H312+H332-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

EINECSNumber : 422-210-5

TSCA : No

Recommended Storage : Ambient temperatures

Documents