missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Propylboronic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1996506

 View more versions of this product

Catalog No. AAL1996506



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Propylboronic acid
105°C to 107°C
MFCD01074564
1731884
n-propylboronic acid, 1-propaneboronic acid, propaneboronic acid, boronic acid, propyl, 1-propylboronic acid, 1-propaneboronic, n-propylboronc acd, propyl boronic acid, pubchem6019
CCCB(O)O
87.91
87.91
17745-45-8
C3H9BO2
5 g
Air sensitive; Hygroscopic
JAQOMSTTXPGKTN-UHFFFAOYSA-N
propylboronic acid
351065
98%

Chemical Identifiers

17745-45-8
87.91
JAQOMSTTXPGKTN-UHFFFAOYSA-N
351065
CCCB(O)O
C3H9BO2
MFCD01074564
n-propylboronic acid, 1-propaneboronic acid, propaneboronic acid, boronic acid, propyl, 1-propylboronic acid, 1-propaneboronic, n-propylboronc acd, propyl boronic acid, pubchem6019
propylboronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only