Please login to your online account to display your discounted pricing

1-Propyl-4-piperidone, 98%, ACROS Organics™

$42.69 - $42.69

Chemical Identifiers

Chemical Name or Material 1-Propyl-4-piperidone
CAS 23133-37-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H15NO
Formula Weight 141.21
MDL Number MFCD00010433
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC134150050

  SDS
thermo fisher scientific
134150050
Each for $42.69

AC134151000

  SDS
thermo fisher scientific
134151000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 141.21
Physical Form Liquid
Quantity 5g
Color Clear yellow
Density 0.9360g/mL
CAS Max % 100.0
Chemical Name or Material 1-Propyl-4-piperidone
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C8H15NO
CAS 23133-37-1
MDL Number MFCD00010433
CAS Min % 97.5
Storage Information Gains color during storage. Product may gradually polymerize during storage
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.