missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Propanol, for spectroscopy ACS, ACROS Organics™

Manufacturer:  Acros Organics AC434360025

 View more versions of this product

Catalog No. AC434360025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

For UV/VIS spectroscopy
Specifications

Specifications

1-Propanol
15°C
Glass bottle
0.001% max.
0.2% max.
Undesignated
2.5L
71-23-8
CH3CH2CH2OH
Solubility in water: soluble. Other solubilities: miscible with alcohol and ether, soluble in many organic solvents
CCCO
60.096
CHEBI:28831
Liquid
ACS
0.8000g/mL
Authentic
1.3840 to 1.3860
0.8
97.0°C
-127.0°C
For Spectroscopy
C3H8O
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
BDERNNFJNOPAEC-UHFFFAOYSA-N
propan-1-ol
1031
60.1
≥99.5%

Chemical Identifiers

71-23-8
60.096
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
CHEBI:28831
CCCO
C3H8O
BDERNNFJNOPAEC-UHFFFAOYSA-N
1031
propan-1-ol
Safety and Handling

Safety and Handling

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents