missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Propanol, Spectrophotometric Grade, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 22933K7

 View more versions of this product

Catalog No. AA22933K7



Specifications

Specifications

1-Propanol
Sprectrophotometric
99%
MFCD00002941
BDERNNFJNOPAEC-UHFFFAOYSA-N
propan-1-ol
1031
60.1
Alcohol-like
Undesignated
71-23-8
Clear liquid
C3H8O
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
CCCO
60.096
CHEBI:28831
Liquid
13.3 hPa (10mm Hg) at 20°C
4L

Chemical Identifiers

71-23-8
60.096
BDERNNFJNOPAEC-UHFFFAOYSA-N
1031
propan-1-ol
C3H8O
MFCD00002941
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
CHEBI:28831
CCCO
Safety and Handling

Safety and Handling

H225-H318-H336: Highly flammable liquid and vapour. Causes serious eye damage. May cause drowsiness or dizziness.

DOTInformation : DOT Class: 3; Packing Group: II

EINECSNumber : 200-746-9

SDS Documents