Please login to your online account to display your discounted pricing

1-Propanol, Spectrophotometric Grade, 99%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material 1-Propanol
CAS 71-23-8
Purity Grade Spectrophotometric grade
Physical Form Liquid
Assay Percent Range 99%
Molecular Formula CH3CH2CH2OH
Formula Weight 60.1
MDL Number MFCD00002941
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA22933K2

  SDS
alfa aesar
22933K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA22933K7

  SDS
alfa aesar
22933K7
4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 60.1
Purity Grade Spectrophotometric grade
Quantity 1L
Color Clear
DOT Information DOT Class: 3; Packing Group: II
Chemical Name or Material 1-Propanol
MDL Number MFCD00002941
EINECS Number 200-746-9
Physical Form Liquid
CAS 71-23-8
Health Hazard 1 H225-H318-H336: Highly flammable liquid and vapour. Causes serious eye damage. May cause drowsiness or dizziness.
Odor Alcohol-like
Molecular Formula CH3CH2CH2OH
Assay Percent Range 99%
Appearance Clear liquid
Vapor Pressure 13.3 hPa (10mm Hg) at 20°C
Safety and Handling

H225-H318-H336: Highly flammable liquid and vapour. Causes serious eye damage. May cause drowsiness or dizziness.