Learn More
1-Propanol, specified according to the requirements of Ph.Eur., Thermo Scientific™
Supplier: Thermo Scientific Chemicals 446270010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-Propanol | |
passes test | |
0.8000g/mL | |
15°C | |
Glass bottle | |
1.3840 to 1.3860 | |
MFCD00002941 | |
0.8 | |
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
propan-1-ol | |
1031 | |
CHEBI:28831 | |
≥99.7% (GC) |
71-23-8 | |
0.01 max. at 310nm, 0.02 max. at 290nm, 0.03 max. at 270nm, 0.1 max. at 250nm, 0.3 max. at 230nm | |
97.0°C | |
Authentic | |
C3H8O | |
CH3CH2CH2OH | |
1L | |
15,7955 | |
Solubility in water: soluble. Other solubilities: miscible with alcohol and ether, soluble in many organic solvents | |
CCCO | |
60.10 | |
2.2 mPa.s (20°C) | |
60.1 | |
0.3 (Total impurities) max |
Chemical Identifiers
71-23-8 | |
60.10 | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
1031 | |
propan-1-ol |
C3H8O | |
MFCD00002941 | |
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
CHEBI:28831 | |
CCCO |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness.
Causes serious eye damage.
Highly flammable liquid and vapor.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
GHS Signal Word: Danger
EINECSNumber : 200-746-9
RUO â Research Use Only