missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Propanol, puriss. p.a., ACS Reagent, ≥99.5% (GC), Honeywell™ Riedel-de Haën™
Puriss. p.a., Reag. Ph. Eur.,≥99.5% (GC)
Supplier: Honeywell Chemicals 335384X25L
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
1-Propanol | |
71-23-8 | |
0.804g/mL at 25°C | |
4 x 2.5 L | |
22°C (71.6°F) | |
Alcohol-like | |
n20/D 1.384 | |
CH3CH2CH2OH | |
14.9mmHg (20°C), 10mmHg (147°C) | |
1098242 | |
Soluble in water and most organic solvents | |
CCCO | |
60.10 | |
CHEBI:28831 | |
≥99.5% (GC) |
-127°C | |
Colorless | |
97°C | |
0.137 | |
700°F | |
Poly Bottle | |
C3H8O | |
MFCD00002941 | |
UN1274 | |
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
propan-1-ol | |
1031 | |
60.1g/mol |
Chemical Identifiers
71-23-8 | |
60.10 | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
1031 | |
propan-1-ol |
C3H8O | |
MFCD00002941 | |
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
CHEBI:28831 | |
CCCO |
Safety and Handling
P210-P261-P280-P305 + P351 + P338
DOTInformation : Class 3 - PG 2 - n-Propanol
RTECSNumber : 200-746-9