Learn More
1-Propanol, HPLC Grade, 99% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 022932M1
Description
1-Propanol is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Propanol | |
°C to 127°C | |
97°C | |
99% min. | |
C3H8O | |
MFCD00002941 | |
1098242 | |
Miscible with water,alcohols,ketones,aldehydes,ethers,glycols,aromatic and aliphatic hydrocarbons. | |
CCCO | |
60.10 | |
CHEBI:28831 | |
13.3 hPa (10mm Hg) at 20°C | |
Liquid |
71-23-8 | |
0.804 | |
15°C (59°F) | |
Alcohol-like | |
1.385 | |
4 x 1 L | |
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
propan-1-ol | |
1031 | |
60.1 | |
HPLC |
Chemical Identifiers
71-23-8 | |
60.10 | |
BDERNNFJNOPAEC-UHFFFAOYSA-N | |
1031 | |
propan-1-ol |
C3H8O | |
MFCD00002941 | |
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
CHEBI:28831 | |
CCCO |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P370+P378q-P501c
H225-H318-H336
DOTInformation : DOT Class: 3; Packing Group: II
EINECSNumber : 200-746-9
RTECSNumber : UH8225000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only