Please login to your online account to display your discounted pricing

1-Propanol, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 4L
Packaging Narrow Mouth Amber Glass Bottle

Manufacturer: avantor performance  908603

 View more versions of this product

Catalog No. 02002051

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 60.1
Chemical Name or Material 1-Propanol
CAS 71-23-8
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.80kg
Purity Grade BAKER ANALYZED™ Reagent
Flash Point 23.00°C
Quantity 4L
Molecular Formula CH3CH2CH2OH

Specifications

Chemical Name or Material 1-Propanol
CAS 71-23-8
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula CH3CH2CH2OH
Formula Weight 60.1