missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Riedel-de Haen™ 1-Propanol, ACS Reagent, ≥99.5%, Honeywell™ Riedel-de Haën™

ACS Reagent,≥99.5%

$133.30 - $133.30

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60046796 View Documents Honeywell Riedel-de Haen™
4028931L
1L Glass Bottle Each for $133.30
 
Description

Description

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Specifications

Chemical Identifiers

71-23-8
60.096
BDERNNFJNOPAEC-UHFFFAOYSA-N
1031
propan-1-ol
C3H8O
MFCD00002941
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
CHEBI:28831
CCCO

Specifications

71-23-8
CH3CH2CH2OH
UN1274
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
CCCO
60.096
CHEBI:28831
≥99.5%
0.137
Glass Bottle
14.9mmHg (20°C), 10mmHg (147°C)
22°C (71.6°F)
97°C
-127°C
1-Propanol
C3H8O
MFCD00002941
1098242
BDERNNFJNOPAEC-UHFFFAOYSA-N
propan-1-ol
1031
60.1g/mol
0.804g/mL at 25°C
700°F
n20/D 1.384
2.1 (vs air)
Liquid
Undesignated
1L
Safety and Handling

Safety and Handling

P210-P261-P280-P305 + P351 + P338

DOTInformation : class 3 - PG 2 - n-Propanol

RTECSNumber : 200-746-9