missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Propanol, ACS, >99.5%, Alfa Aesar

Manufacturer:  Alfa Aesar 43848M6

 View more versions of this product

Catalog No. AA43848M6



Specifications

Specifications

1-Propanol
71-23-8
C3H8O
1-hydroxypropane
CCCO
60.096
CHEBI:28831
Liquid
824°C (1515°F)
10mm Hg at 14°C
4 x 4L
In ACS analysis
>99.5%
MFCD00002941
BDERNNFJNOPAEC-UHFFFAOYSA-N
propan-1-ol
1031
60.1
ACS
Alcohol-like
Colorless

Chemical Identifiers

71-23-8
60.096
BDERNNFJNOPAEC-UHFFFAOYSA-N
1031
propan-1-ol
C3H8O
MFCD00002941
1-hydroxypropane
CHEBI:28831
CCCO
Safety and Handling

Safety and Handling

H225-H318-H336: Highly flammable liquid and vapour. Causes serious eye damage. May cause drowsiness or dizziness.

DOTInformation : DOT Class: 3, Packaging Group: II

Documents