missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Propanol, 99+%, Extra Pure, ACROS Organics™

Manufacturer:  Acros Organics 149480010

 View more versions of this product

Catalog No. AC149480010



Specifications

Specifications

1-Propanol
99.0
Vapor Pressure: 25mbar at 20°C
99+%
CH3CH2CH2OH
01, IV, 1413
4-01-00-01413 beilstein handbook reference, propyl alcohol normal reagent grade acs, 1-propanol uv hplc spectroscopic, policosanol from rice wax oryza sp., n-propanol or propyl alcohol normal, 1-propanol spectrophotometric grade, 1-propanol saj first grade, epa pesticide chemical code 047502, propyl alcohol fragrance grade, 1-propanol acs reagent
CCCO
60.096
CHEBI:28831
Liquid
0.8000g/mL
Glass Bottle
0.005% max.
Undesignated
71-23-8
100.0
Extra Pure
C3H8O
MFCD00002941
15, 7955
BDERNNFJNOPAEC-UHFFFAOYSA-N
propan-1-ol
1031
60.1
≥99.0%
Authentic
1.3840 to 1.3860 (20°C, 589nm)
0.5% max.
1L

Chemical Identifiers

71-23-8
60.096
BDERNNFJNOPAEC-UHFFFAOYSA-N
1031
propan-1-ol
C3H8O
MFCD00002941
4-01-00-01413 beilstein handbook reference, propyl alcohol normal reagent grade acs, 1-propanol uv hplc spectroscopic, policosanol from rice wax oryza sp., n-propanol or propyl alcohol normal, 1-propanol spectrophotometric grade, 1-propanol saj first grade, epa pesticide chemical code 047502, propyl alcohol fragrance grade, 1-propanol acs reagent
CHEBI:28831
CCCO
Safety and Handling

Safety and Handling

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents