missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Propanethiol, 98%

Manufacturer:  Alfa Aesar™ 43420AP

 View more versions of this product

Catalog No. AA43420AP



Description

Description

  • Packaged under Argon
Specifications

Specifications

1-Propanethiol
−20°C (4°F)
1.4380
67°C to 68°C
-113°C
107-03-9
CH3CH2CH2SH
UN2402
SUVIGLJNEAMWEG-UHFFFAOYSA-N
propane-1-thiol
7848
76.16
98%
0.8410g/mL
Stench
Air Sensitive
Colorless
500mL
C3H8S
MFCD00004900
1-propanethiol, propanethiol, n-propylmercaptan, propyl mercaptan, n-propyl mercaptan, n-propylthiol, 1-propylmercaptan, propylthiol, 1-mercaptopropane, 1-propyl mercaptan
CCCS
76.157
CHEBI:8473
Liquid

Chemical Identifiers

107-03-9
76.157
SUVIGLJNEAMWEG-UHFFFAOYSA-N
7848
propane-1-thiol
C3H8S
MFCD00004900
1-propanethiol, propanethiol, n-propylmercaptan, propyl mercaptan, n-propyl mercaptan, n-propylthiol, 1-propylmercaptan, propylthiol, 1-mercaptopropane, 1-propyl mercaptan
CHEBI:8473
CCCS
Safety and Handling

Safety and Handling

GHS H Statement
H225-H318-H302-H335-H315
Highly flammable liquid and vapor.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P210-P261-P280-P273-P305+P351+P338-P403
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store in a well-ventilated place.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PROPANETHIOLS

RTECSNumber : TZ7300000

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

Documents