Please login to your online account to display your discounted pricing

1-Propanesulfonic acid, sodium salt, 98%, Acros Organics


Quantity 5g

Manufacturer: thermo fisher scientific  338450050

 View more versions of this product

Catalog No. AC338450050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 146.14
Physical Form Fine Crystalline Powder
Quantity 5g
Color White
TSCA TSCA
EINECS Number 238-564-7
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS Min % 97.5
Synonym Sodium 1-propanesulfonate
Chemical Name or Material 1-Propanesulfonic acid, sodium salt
MDL Number MFCD00062546
Assay Percent Range 98%
Name Note 98%
CAS 14533-63-2
Molecular Formula C3H7NaO3S

Specifications

Chemical Name or Material 1-Propanesulfonic acid, sodium salt
Name Note 98%
CAS 14533-63-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C3H7NaO3S
Formula Weight 146.14
MDL Number MFCD00062546
Synonym Sodium 1-propanesulfonate
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.