missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Propanesulfonic acid, sodium salt, 98%, Acros Organics

$196.60 - $196.60

Chemical Identifiers

CAS 14533-63-2
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C3H7NaO3S
Formula Weight 146.14
MDL Number MFCD00062546
Synonym Sodium 1-propanesulfonate
Chemical Name or Material 1-Propanesulfonic acid, sodium salt, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC338450050 SDS Thermo Fisher Scientific
338450050
5g Each for $196.60
Specifications Description & Specifications

Chemical Identifiers

14533-63-2
100.0
C3H7NaO3S
MFCD00062546
1-Propanesulfonic acid, sodium salt, 98%
97.5
98%
146.14
Sodium 1-propanesulfonate
≥97.5%

Specifications

14533-63-2
100.0
Fine Crystalline Powder
C3H7NaO3S
MFCD00062546
White
1-Propanesulfonic acid, sodium salt, 98%
97.5
Authentic
98%
146.14
Sodium 1-propanesulfonate
5g
≥97.5%