Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Piperonylpiperazine, 97%

Chemical Identifiers

Chemical Name or Material 1-Piperonylpiperazine
CAS 32231-06-4
Molecular Formula C12H16N2O2
Formula Weight 220.27
MDL Number MFCD00005834
UN Number UN3259
Beilstein 885038
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1562506

  SDS
alfa aesar™
L1562506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1562514

  SDS
alfa aesar™
L1562514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 220.27
Chemical Name or Material 1-Piperonylpiperazine
Assay 97%
Molecular Formula C12H16N2O2
MDL Number MFCD00005834
UN Number UN3259
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 147° to 149°C (2mmHg)
CAS 32231-06-4
Beilstein 885038
Melting Point 40° to 45°C
Quantity 5g
Flash Point >110°C (230°F)
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
RTECS Number TM1550000
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.