Please login to your online account to display your discounted pricing

1-Piperidino-1-cyclohexene 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  393270250

 View more versions of this product

Catalog No. AC393270250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5130 to 1.5150 (20°C, 589nm)
Density 0.98g/mL
Color Clear yellow
Quantity 25g
Chemical Name or Material 1-Piperidino-1-cyclohexene
CAS 395106
CAS Min % 94.0
CAS Max % 100.0
Physical Form Clear yellow liquid
Assay 94% min.
Assay Percent Range 95%
Molecular Formula C11H19N
Formula Weight 165.28g/mol
MDL Number MFCD00014643
Beilstein 20, V,2, 48
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-Piperidino-1-cyclohexene
CAS 395106
CAS Min % 94.0
CAS Max % 100.0
Assay 94% min.
Assay Percent Range 95%
Molecular Formula C11H19N
Formula Weight 165.28g/mol
MDL Number MFCD00014643
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents