Please login to your online account to display your discounted pricing

1-Piperidino-1-cyclohexene 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Piperidino-1-cyclohexene
CAS 395106
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C11H19N
Formula Weight 165.28
MDL Number MFCD00014643
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC393270050

  SDS
thermo fisher scientific
AC393270050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Liquid
Formula Weight 165.28
Quantity 5g
Color Clear yellow
Density 0.98g/mL
CAS 395106
CAS Max % 100.0
CAS Min % 94.0
Beilstein 20, V,2, 48
Packaging Glass bottle
Chemical Name or Material 1-Piperidino-1-cyclohexene
Assay Percent Range 95%
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00014643
Infrared Spectrum Authentic
Refractive Index 1.5130 to 1.5150 (20°C, 589nm)
Molecular Formula C11H19N
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.