Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Piperidinepropionitrile, 98+%

Chemical Identifiers

Chemical Name or Material 1-Piperidinepropionitrile
CAS 3088-41-3
Molecular Formula C8H14N2
Formula Weight 138.21
MDL Number MFCD00006511
UN Number UN3276
Beilstein 107858
Synonym 1-(2-Cyanoethyl)piperidine; 3-(1-Piperidino)propionitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0146918

  SDS
alfa aesar™
L0146918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0146930

  SDS
alfa aesar™
L0146930
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 138.21
Assay ≥98%
MDL Number MFCD00006511
TSCA TSCA
Molecular Formula C8H14N2
CAS 3088-41-3
UN Number UN3276
Beilstein 107858
Synonym 1-(2-Cyanoethyl)piperidine; 3-(1-Piperidino)propionitrile
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 102°C (215°F)
Boiling Point 110° to 111°C (16mmHg)
Chemical Name or Material 1-Piperidinepropionitrile
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
RTECS Number TN3714000
Quantity 50g
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.