Please login to your online account to display your discounted pricing

1-Piperidinepropionic acid, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Piperidinepropionic acid
CAS 26371-07-3
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95.0%
Assay Percent Range 96%
Molecular Formula C8H15NO2
Formula Weight 157.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC440050250

  SDS
thermo fisher scientific
AC440050250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC440051000

  SDS
thermo fisher scientific
AC440051000
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥95.0%
Formula Weight 157.21
Quantity 25g
Color White to pale brown
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material 1-Piperidinepropionic acid
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Assay Percent Range 96%
Molecular Formula C8H15NO2
CAS 26371-07-3
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.