Please login to your online account to display your discounted pricing

1-Piperazinecarboxaldehyde, 90%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Piperazinecarboxaldehyde
CAS 7755-92-2
CAS Min % 88.0
CAS Max % 100.0
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C5H10N2O
Formula Weight 114.15
Synonym 1-Formylpiperazine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC388490250

  SDS
thermo fisher scientific
AC388490250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC388491000

  SDS
thermo fisher scientific
AC388491000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥88%
Formula Weight 114.15
Physical Form Liquid
Quantity 25g
Color Clear colorless to yellow
Density 1.1g/mL
CAS Max % 100.0
CAS Min % 88.0
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C5H10N2O
Packaging Glass bottle
Refractive Index 1.5090 to 1.5130 (20°C, 589nm)
Synonym 1-Formylpiperazine
Health Hazard 1 GHS Signal Word: Danger
CAS 7755-92-2
Chemical Name or Material 1-Piperazinecarboxaldehyde
Assay Percent Range 90%
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.